cis-3-Amino-2,2-dimethylcyclobutanol hydrochloride

Lieferant: Apollo Scientific

OR304340-1G OR304340-250MG OR304340-500MG
APOSOR304340-1GEA 1900 EUR
APOSOR304340-1G APOSOR304340-250MG APOSOR304340-500MG
cis-3-Amino-2,2-dimethylcyclobutanol hydrochloride
cis-3-Amino-2,2-dimethylcyclobutanol hydrochloride
Formel: C₆H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27956964
CAS-Nummer: 1434141-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR