3-Hydroxycyclobutane-1-carbonitrile

Lieferant: Apollo Scientific

OR304343-1G OR304343-5G
APOSOR304343-1GEA 237 EUR
APOSOR304343-1G APOSOR304343-5G
3-Hydroxycyclobutane-1-carbonitrile
3-Hydroxycyclobutane-1-carbonitrile
Formel: C₅H₇NO
Molecular Weight: 97.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13659429
CAS-Nummer: 20249-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR