3-(Dimethylamino)cyclobutan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR304360-1G
OR304360-250MG
APOSOR304360-1GEA
1560
EUR
APOSOR304360-1G
APOSOR304360-250MG
3-(Dimethylamino)cyclobutan-1-ol
3-(Dimethylamino)cyclobutan-1-ol
Formel:
C₆H₁₃NO Molecular Weight: 115,174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24530651 CAS-Nummer: 18126-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...