1-(3-Hydroxypropyl)cyclobutan-1-ol

Lieferant: Apollo Scientific

OR304362-1G OR304362-5G
APOSOR304362-1GEA 0 EUR
APOSOR304362-1G APOSOR304362-5G
1-(3-Hydroxypropyl)cyclobutan-1-ol
1-(3-Hydroxypropyl)cyclobutan-1-ol
Formel: C₇H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28098637
CAS-Nummer: 1584139-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR