trans-3-(Benzyloxy)cyclobutanol

Lieferant: Apollo Scientific

OR304363-1G OR304363-250MG
APOSOR304363-1GEA 473 EUR
APOSOR304363-1G APOSOR304363-250MG
trans-3-(Benzyloxy)cyclobutanol
trans-3-(Benzyloxy)cyclobutanol
Formel: C₁₁H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22397416
CAS-Nummer: 1383813-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR