1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid

Lieferant: Apollo Scientific

OR304382-1G OR304382-5G
APOSOR304382-1GEA 589 EUR
APOSOR304382-1G APOSOR304382-5G
1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid
1-(Prop-2-en-1-yl)cyclobutane-1-carboxylic acid
Formel: C₈H₁₂O₂
Molecular Weight: 140.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19229282
CAS-Nummer: 1193397-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR