(5-Methylpyridazin-3-yl)methanamine hydrochloride

Lieferant: Apollo Scientific

OR304388-100MG OR304388-250MG
APOSOR304388-100MGEA 567 EUR
APOSOR304388-100MG APOSOR304388-250MG
(5-Methylpyridazin-3-yl)methanamine hydrochloride
(5-Methylpyridazin-3-yl)methanamine hydrochloride
Formel: C₆H₉N₃·ClH
Molecular Weight: 159,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037452
CAS-Nummer: 1788054-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR