5-Chloropyridazin-3-amine

Lieferant: Apollo Scientific

OR304389-1G OR304389-250MG
APOSOR304389-1GEA 496 EUR
APOSOR304389-1G APOSOR304389-250MG
5-Chloropyridazin-3-amine
5-Chloropyridazin-3-amine
Formel: C₄H₄ClN₃
Molecular Weight: 129,548 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19214495
CAS-Nummer: 1314978-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR