(5-Methylpyridazin-3-yl)methanol

Lieferant: Apollo Scientific

OR304392-1G OR304392-250MG
APOSOR304392-1GEA 250 EUR
APOSOR304392-1G APOSOR304392-250MG
(5-Methylpyridazin-3-yl)methanol
(5-Methylpyridazin-3-yl)methanol
Formel: C₆H₈N₂O
Molecular Weight: 124,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037453
CAS-Nummer: 1788044-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR