5-Methylpyridazine-4-carbonitrile

Lieferant: Apollo Scientific

OR304393-1G OR304393-250MG OR304393-500MG
APOSOR304393-1GEA 1270 EUR
APOSOR304393-1G APOSOR304393-250MG APOSOR304393-500MG
5-Methylpyridazine-4-carbonitrile
5-Methylpyridazine-4-carbonitrile
Formel: C₆H₅N₃
Molecular Weight: 119.124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30471638
CAS-Nummer: 2007921-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR