3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride

Lieferant: Apollo Scientific

OR304398-1G OR304398-5G
APOSOR304398-1GEA 700 EUR
APOSOR304398-1G APOSOR304398-5G
3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride
3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride
Formel: C₁₀H₂₀N₂·2ClH
Molecular Weight: 241.201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185054
CAS-Nummer: 13323-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR