2,6-Diazaspiro[3.5]nonan-1-one

Lieferant: Apollo Scientific

OR304404-250MG OR304404-500MG
APOSOR304404-250MGEA 705 EUR
APOSOR304404-250MG APOSOR304404-500MG
2,6-Diazaspiro[3.5]nonan-1-one
2,6-Diazaspiro[3.5]nonan-1-one
Formel: C₇H₁₂N₂O
Molecular Weight: 140,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23378266
CAS-Nummer: 1422062-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR