Spiro[3.5]nonan-2-one

Lieferant: Apollo Scientific

OR304405-1G OR304405-5G
APOSOR304405-1GEA 433 EUR
APOSOR304405-1G APOSOR304405-5G
Spiro[3.5]nonan-2-one
Spiro[3.5]nonan-2-one
Formel: C₉H₁₄O
Molecular Weight: 138,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483741
CAS-Nummer: 29800-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR