Spiro[3.3]heptan-2-ol

Lieferant: Apollo Scientific
Achtung

OR304421-1G OR304421-500MG
APOSOR304421-1GEA 1270 EUR
APOSOR304421-1G APOSOR304421-500MG
Spiro[3.3]heptan-2-ol
Spiro[3.3]heptan-2-ol
Formel: C₇H₁₂O
Molecular Weight: 112.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23884142
CAS-Nummer: 1502422-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR