2-Thiaspiro[3.3]heptan-6-amine

Lieferant: Apollo Scientific

OR304427-100MG OR304427-250MG OR304427-500MG
APOSOR304427-100MGEA 601 EUR
APOSOR304427-100MG APOSOR304427-250MG APOSOR304427-500MG
2-Thiaspiro[3.3]heptan-6-amine
2-Thiaspiro[3.3]heptan-6-amine
Formel: C₆H₁₁NS
Molecular Weight: 129.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566239
CAS-Nummer: 1363381-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR