6-Oxaspiro[4.5]decan-9-one

Lieferant: Apollo Scientific

OR304430-1G OR304430-5G
APOSOR304430-1GEA 678 EUR
APOSOR304430-1G APOSOR304430-5G
6-Oxaspiro[4.5]decan-9-one
6-Oxaspiro[4.5]decan-9-one
Formel: C₉H₁₄O₂
Molecular Weight: 154.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19651152
CAS-Nummer: 855398-57-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR