tert-Butyl (3,4-diaminophenyl)carbamate

Lieferant: Apollo Scientific
Achtung

OR305000-1G OR305000-250MG
APOSOR305000-1GEA 763 EUR
APOSOR305000-1G APOSOR305000-250MG
tert-Butyl (3,4-diaminophenyl)carbamate
tert-Butyl (3,4-diaminophenyl)carbamate
Formel: C₁₁H₁₇N₃O₂
Molecular Weight: 223,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13182465
CAS-Nummer: 937372-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR