Methyl 2-iodo-6-methylbenzoate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-iodo-o-toluate, 3-Iodo-2-(methoxycarbonyl)toluene

OR305011-1G OR305011-250MG OR305011-5G
APOSOR305011-1GEA 29 EUR
APOSOR305011-1G APOSOR305011-250MG APOSOR305011-5G
Methyl 2-iodo-6-methylbenzoate 97%
Methyl 2-iodo-6-methylbenzoate
Formel: C₉H₉IO₂
Molecular Weight: 276,071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061917
CAS-Nummer: 103440-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR