6-Chloro-1H-indazole-3-carboxaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-1H-indazole-3-carbaldehyde

OR305084-1G OR305084-250MG OR305084-5G
APOSOR305084-1GEA 56.7 EUR
APOSOR305084-1G APOSOR305084-250MG APOSOR305084-5G
6-Chloro-1H-indazole-3-carboxaldehyde 95%
6-Chloro-1H-indazole-3-carboxaldehyde
Formel: C₈H₅ClN₂O
Molecular Weight: 180,591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781580
CAS-Nummer: 885521-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR