Methyl 5-bromo-1H-indazole-6-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-(methoxycarbonyl)-1H-indazole

OR305115-1G OR305115-250MG
APOSOR305115-1GEA 42.2 EUR
APOSOR305115-1G APOSOR305115-250MG
Methyl 5-bromo-1H-indazole-6-carboxylate 97%
Methyl 5-bromo-1H-indazole-6-carboxylate
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255.068 g/mol
Schmelzpunkt: 148…153 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09749941
CAS-Nummer: 1000342-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR