3H-Imidazo[4,5-b]pyridine-6-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Carboxy-3H-imidazo[4,5-b]pyridine
OR305191-1G
OR305191-250MG
APOSOR305191-1GEA
299
EUR
APOSOR305191-1G
APOSOR305191-250MG
3H-Imidazo[4,5-b]pyridine-6-carboxylic acid
3H-Imidazo[4,5-b]pyridine-6-carboxylic acid
Formel:
C₇H₅N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06254751 CAS-Nummer: 24638-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...