Methyl 1-oxo-2,3-dihydro-1H-isoindole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-oxoisoindoline-5-carboxylate

OR305278-1G OR305278-250MG
APOSOR305278-1GEA 386 EUR
APOSOR305278-1G APOSOR305278-250MG
Methyl 1-oxo-2,3-dihydro-1H-isoindole-5-carboxylate
Methyl 1-oxo-2,3-dihydro-1H-isoindole-5-carboxylate
Formel: C₁₀H₉NO₃
Molecular Weight: 191,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177815
CAS-Nummer: 926307-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR