3-Oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Oxoisoindoline-4-carboxylic acid, 3-Oxo-4-isoindolinecarboxylic acid
OR305297-250MG
APOSOR305297-250MGEA
120
EUR
APOSOR305297-250MG
3-Oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid
3-Oxo-2,3-dihydro-1H-isoindole-4-carboxylic acid
Formel:
C₉H₇NO₃ Molecular Weight: 177,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11506114 CAS-Nummer: 935269-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...