7-Bromo-2,3-dihydro-1H-isoindol-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-2,3-dihydro-isoindol-1-one

OR305302-250MG OR305302-1G
APOSOR305302-250MGEA 24.8 EUR
APOSOR305302-250MG APOSOR305302-1G
7-Bromo-2,3-dihydro-1H-isoindol-1-one 95%
7-Bromo-2,3-dihydro-1H-isoindol-1-one
Formel: C₈H₆BrNO
Molecular Weight: 212,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848600
CAS-Nummer: 200049-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR