3-(Hydroxymethyl)isoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: Isoxazol-3-ylmethanol

OR305308-1G OR305308-250MG
APOSOR305308-1GEA 104 EUR
APOSOR305308-1G APOSOR305308-250MG
3-(Hydroxymethyl)isoxazole
3-(Hydroxymethyl)isoxazole
Formel: C₄H₅NO₂
Molecular Weight: 99,088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778413
CAS-Nummer: 89102-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR