3-(Sulphanylmethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Pyridinemethanethiol, 3-(Mercaptomethyl)pyridine, (Pyridin-3-yl)methanethiol
OR305406-1G
OR305406-250MG
APOSOR305406-1GEA
749
EUR
APOSOR305406-1G
APOSOR305406-250MG
3-(Sulphanylmethyl)pyridine 95%
3-(Sulphanylmethyl)pyridine
Formel:
C₆H₇NS Molecular Weight: 125,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01691719 CAS-Nummer: 17617-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...