(6-Aminopyridin-3-yl)acetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(carboxymethyl)pyridine, 2-(6-Aminopyridin-3-yl)ethanoic acid

OR305409-250MG
APOSOR305409-250MGEA 760 EUR
APOSOR305409-250MG
(6-Aminopyridin-3-yl)acetic acid 95%
(6-Aminopyridin-3-yl)acetic acid
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09757480
CAS-Nummer: 39658-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR