4-Hydroxy-2-methoxypyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxypyrimidin-4-ol, 2-Methoxypyrimidin-4(1H)-one, 2-Methoxypyrimidin-4(3H)-one
OR30542-1G
OR30542-250MG
APOSOR30542-1GEA
78.3
EUR
APOSOR30542-1G
APOSOR30542-250MG
4-Hydroxy-2-methoxypyrimidine 97%
4-Hydroxy-2-methoxypyrimidine
Formel:
C₅H₆N₂O₂ Molecular Weight: 126,113 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00128202 CAS-Nummer: 25902-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...