Imidazo[1,2-a]pyridin-7-ol

Lieferant: Apollo Scientific

OR305451-250MG OR305451-100MG OR305451-1G
APOSOR305451-250MGEA 200 EUR
APOSOR305451-250MG APOSOR305451-100MG APOSOR305451-1G
Imidazo[1,2-a]pyridin-7-ol
Imidazo[1,2-a]pyridin-7-ol
Formel: C₇H₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13176749
CAS-Nummer: 896139-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR