6-Bromo-5-chloro-1H-indazole

Lieferant: Apollo Scientific

OR305514-250MG OR305514-1G OR305514-5G
APOSOR305514-250MGEA 114 EUR
APOSOR305514-250MG APOSOR305514-1G APOSOR305514-5G
6-Bromo-5-chloro-1H-indazole
6-Bromo-5-chloro-1H-indazole
Formel: C₇H₄BrClN₂
Molecular Weight: 231.477 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689251
CAS-Nummer: 1305208-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR