6-(Hydroxymethyl)isoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: (Isoquinolin-6-yl)methanol, 6-(Hydroxymethyl)-2-azanaphthalene, Isoquinolin-6-ylmethanol

OR30565-1G OR30565-250MG
APOSOR30565-1GEA 358 EUR
APOSOR30565-1G APOSOR30565-250MG
6-(Hydroxymethyl)isoquinoline
6-(Hydroxymethyl)isoquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159.185 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11226912
CAS-Nummer: 188861-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR