4-Bromo-1,8-naphthyridine 95%

Lieferant: Apollo Scientific
Achtung

OR30601-1G OR30601-250MG
APOSOR30601-1GEA 938 EUR
APOSOR30601-1G APOSOR30601-250MG
4-Bromo-1,8-naphthyridine 95%
4-Bromo-1,8-naphthyridine
Formel: C₈H₅BrN₂
Molecular Weight: 209,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520860
CAS-Nummer: 54569-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR