8-Bromopyrido[4,3-b]pyrazine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 8-Bromopyrido[3,4-b]pyrazine

OR30605-1G OR30605-250MG
APOSOR30605-1GEA 372 EUR
APOSOR30605-1G APOSOR30605-250MG
8-Bromopyrido[4,3-b]pyrazine
8-Bromopyrido[4,3-b]pyrazine
Formel: C₇H₄BrN₃
Molecular Weight: 210,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834590
CAS-Nummer: 929074-45-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR