3-Cyclopropylisonicotinic acid

Lieferant: Apollo Scientific

Synonyme: 3-Cyclopropylpyridine-4-carboxylic acid

OR306658-1G OR306658-250MG
APOSOR306658-1GEA 575 EUR
APOSOR306658-1G APOSOR306658-250MG
3-Cyclopropylisonicotinic acid
3-Cyclopropylisonicotinic acid
Formel: C₉H₉NO₂
Molecular Weight: 163.173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18261166
CAS-Nummer: 1256785-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR