1H-Indazole-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H-Indazole-3-carbaldehyde

OR30687-1G OR30687-5G OR30687-10G
APOSOR30687-1GEA 29 EUR
APOSOR30687-1G APOSOR30687-5G APOSOR30687-10G
1H-Indazole-3-carboxaldehyde
1H-Indazole-3-carboxaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146.146 g/mol
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972468
CAS-Nummer: 5235-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR