2-Hydrazino-6-cyanopyridine

Lieferant: Apollo Scientific
Achtung

OR307017-1G OR307017-250MG
APOSOR307017-1GEA 551 EUR
APOSOR307017-1G APOSOR307017-250MG
2-Hydrazino-6-cyanopyridine
2-Hydrazino-6-cyanopyridine
Formel: C₆H₆N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD20367091
CAS-Nummer: 1339085-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR