3-Iodo-4-nitro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307163-1G OR307163-250MG
APOSOR307163-1GEA 279 EUR
APOSOR307163-1G APOSOR307163-250MG
3-Iodo-4-nitro-1H-pyrazole
3-Iodo-4-nitro-1H-pyrazole
Formel: C₃H₂IN₃O₂
Molecular Weight: 238,971 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976491
CAS-Nummer: 1395443-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR