4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-(1-ethoxy-ethyl)-1H-pyrazole

OR307168-1G OR307168-250MG
APOSOR307168-1GEA 136 EUR
APOSOR307168-1G APOSOR307168-250MG
4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole
4-Bromo-1-(1-ethoxyethyl)-1H-pyrazole
Formel: C₇H₁₁BrN₂O
Molecular Weight: 219.079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22543653
CAS-Nummer: 1024120-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR