1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(1-Ethoxy-ethyl)-4-iodo-1H-pyrazole
OR307170-1G
OR307170-25G
OR307170-5G
APOSOR307170-1GEA
43.5
EUR
APOSOR307170-1G
APOSOR307170-25G
APOSOR307170-5G
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole
1-(1-Ethoxyethyl)-4-iodo-1H-pyrazole
Formel:
C₇H₁₁IN₂O Molecular Weight: 266.08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123601 CAS-Nummer: 575452-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...