3-Bromo-1-(1-ethoxyethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307176-1G OR307176-5G
APOSOR307176-1GEA 154 EUR
APOSOR307176-1G APOSOR307176-5G
3-Bromo-1-(1-ethoxyethyl)-1H-pyrazole
3-Bromo-1-(1-ethoxyethyl)-1H-pyrazole
Formel: C₇H₁₁BrN₂O
Molecular Weight: 219.079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976501
CAS-Nummer: 880767-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR