1-Benzofuran-7-ylmethanol

Lieferant: Apollo Scientific
Achtung

OR307184-1G OR307184-250MG
APOSOR307184-1GEA 299 EUR
APOSOR307184-1G APOSOR307184-250MG
1-Benzofuran-7-ylmethanol
1-Benzofuran-7-ylmethanol
Formel: C₉H₈O₂
Molecular Weight: 148.159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441765
CAS-Nummer: 209256-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR