1-Bromo-2-cyclopropylbenzene

Lieferant: Apollo Scientific
Achtung

OR307196-1G OR307196-5G OR307196-25G
APOSOR307196-1GEA 212 EUR
APOSOR307196-1G APOSOR307196-5G APOSOR307196-25G
1-Bromo-2-cyclopropylbenzene
1-Bromo-2-cyclopropylbenzene
Formel: C₉H₉Br
Molecular Weight: 197.072 g/mol
Siedepunkt: 110…113 °C (18 mm Hg)
Lagertemperatur: Gefrierschrank
MDL: MFCD11109720
CAS-Nummer: 57807-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR