1-Chloro-2-cyclopropylbenzene

Lieferant: Apollo Scientific

OR307245-1G OR307245-250MG
APOSOR307245-1GEA 574 EUR
APOSOR307245-1G APOSOR307245-250MG
1-Chloro-2-cyclopropylbenzene
1-Chloro-2-cyclopropylbenzene
Formel: C₉H₉Cl
Molecular Weight: 152,621 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15527175
CAS-Nummer: 10292-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR