2-Amino-5-cyclopropylpyridine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Cyclopropylpyridin-2-amine
OR307258-1G
OR307258-250MG
APOSOR307258-1GEA
246
EUR
APOSOR307258-1G
APOSOR307258-250MG
2-Amino-5-cyclopropylpyridine
2-Amino-5-cyclopropylpyridine
Formel:
C₈H₁₀N₂ Molecular Weight: 134.178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802676 CAS-Nummer: 893738-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...