3,4-Dibromo-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307262-1G OR307262-250MG
APOSOR307262-1GEA 120 EUR
APOSOR307262-1G APOSOR307262-250MG
3,4-Dibromo-1H-pyrazole
3,4-Dibromo-1H-pyrazole
Formel: C₃H₂Br₂N₂
Molecular Weight: 225,869 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159639
CAS-Nummer: 5932-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR