Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR307287-1G OR307287-250MG OR307287-5G
APOSOR307287-1GEA 284 EUR
APOSOR307287-1G APOSOR307287-250MG APOSOR307287-5G
Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate
Ethyl 3,5-dibromo-1H-pyrazole-4-carboxylate
Formel: C₆H₆Br₂N₂O₂
Molecular Weight: 297,932 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22372689
CAS-Nummer: 1017802-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR