4-Methyl-1H-pyrazol-3-amine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR307288-1G
OR307288-250MG
OR307288-5G
APOSOR307288-1GEA
213
EUR
APOSOR307288-1G
APOSOR307288-250MG
APOSOR307288-5G
4-Methyl-1H-pyrazol-3-amine 97%
4-Methyl-1H-pyrazol-3-amine
Formel:
C₄H₇N₃ Molecular Weight: 97.118 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08272068 CAS-Nummer: 64781-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...