Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate 99%

Lieferant: Apollo Scientific
Achtung

OR307299-1G OR307299-250MG OR307299-5G
APOSOR307299-1GEA 265 EUR
APOSOR307299-1G APOSOR307299-250MG APOSOR307299-5G
Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-5-bromo-1H-pyrazole-4-carboxylate
Formel: C₆H₈BrN₃O₂
Molecular Weight: 234.051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28955031
CAS-Nummer: 1352903-61-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR