1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR307301-1G
OR307301-5G
APOSOR307301-1GEA
383
EUR
APOSOR307301-1G
APOSOR307301-5G
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
1-(1-Ethoxyethyl)-3-iodo-1H-pyrazole
Formel:
C₇H₁₁IN₂O Molecular Weight: 266.08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30721063 CAS-Nummer: 1639858-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...