2-Amino-3,6-dimethylquinoline hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3,6-dimethyl-1-azanaphthalene hydrochloride, 3,6-Dimethylquinolin-2-amine hydrochloride
OR307517-1G
OR307517-5G
APOSOR307517-1GEA
293
EUR
APOSOR307517-1G
APOSOR307517-5G
2-Amino-3,6-dimethylquinoline hydrochloride
2-Amino-3,6-dimethylquinoline hydrochloride
Formel:
C₁₁H₁₂N₂·ClH Molecular Weight: 208,687 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10703335 CAS-Nummer: 1170838-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...